Multi-panel drug tests
About this item
These multipanel drugs tests can detect up to 12 drugs simultaneously.
- Clear results in less than three minutes
- Great accuracy, wide range of tests available
- For detecting recent drug use approximately 1 to 8 days
Suitable for on-site screening, safety critical situations, random screening, for cause screening, and monitoring rehabilitation.
AMP: Amphetamines ; BAR: Barbiturates ; BUP: Buprenorphine ; BZO: Benzodiazepines ; CLON: Clonezepam ; COC: Cocaine ; EDDP: Methadone Metabolite ; FEN: Fentanyl ; KET: Ketamine ; MAMP: Methamphetamines ; MDMA: Ecstasy ; MOP: Morphine ; MTD: Methadone ; OPI: Opiates ; OXY: Oxycodone ; PCP: Phencyclidine ; PPX: Propoxyphene ; TCA: Tricyclic Antidepressants ; TRA: Tramadol ; THC: Cannabis.
Certifications: ISO 9001, ISO 13485. IVD in accordance with 98/79/EC.
Packaging: Individually wrapped in a foil pouch, which ensures that the high-quality of the product is retained throughout the shelf-life.
1 Options Available Below
Product Details & Documents
These multipanel drugs tests can detect up to 12 drugs simultaneously.
- Clear results in less than three minutes
- Great accuracy, wide range of tests available
- For detecting recent drug use approximately 1 to 8 days
Suitable for on-site screening, safety critical situations, random screening, for cause screening, and monitoring rehabilitation.
AMP: Amphetamines ; BAR: Barbiturates ; BUP: Buprenorphine ; BZO: Benzodiazepines ; CLON: Clonezepam ; COC: Cocaine ; EDDP: Methadone Metabolite ; FEN: Fentanyl ; KET: Ketamine ; MAMP: Methamphetamines ; MDMA: Ecstasy ; MOP: Morphine ; MTD: Methadone ; OPI: Opiates ; OXY: Oxycodone ; PCP: Phencyclidine ; PPX: Propoxyphene ; TCA: Tricyclic Antidepressants ; TRA: Tramadol ; THC: Cannabis.
Certifications: ISO 9001, ISO 13485. IVD in accordance with 98/79/EC.
Packaging: Individually wrapped in a foil pouch, which ensures that the high-quality of the product is retained throughout the shelf-life.
Product Family Options
Product Information
- TestsAvailabilityPrice
- SUREG10DS2Multi-panel drug testsMulti-panel drug tests(AMP/mAMP/BAR/BZO/COC/MDMA/MTD/OPI/TCA/THC)Log in for availability1 * 25 STSFr. 291.00Specifications:
More
Specifications
Cat. No.SUREG10DS2 - SUREG10DS5300Multi-panel drug testsMulti-panel drug tests(AMP300 /mAMP300/BUP/BZO/BZO200/COC/MDMA300/MOP/MTD/THC)Discontinued-Specifications:
More
Specifications
Cat. No.SUREG10DS5300