ContactUsLinkComponent
Blow Lamp, X 1650
Blow Lamp, X 1650
Catalog # CAMP2000026867
Supplier:  CAMPING GAZ
Blow Lamp, X 1650
Catalog # CAMP2000026867
Supplier:  CAMPING GAZ
Specifications
  • Description:
    Extra fine flame burner X1650
  • Pk:
    1
Specifications
About this item
Extra fine flame burner. Ideal for any precise work such as soldering small components, jewelry or models
  • Easy screw attachment
  • Gas consumption: 30 g/h