
Ammonium tetrafluoroborate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13826-83-0
:
Formula:H₄BF₄N
MDL Number:MFCD06245785
:104.84 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:237-531-4
UN:3260
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Density:1.85 g/cm3 (20 °C)
- pH:3.5 (5 g/l, H2O, 20 °C)
- Storage temperature:+15...+25 °C
Specification Test Results
Frequently Bought Together