Order Entry
Switzerland
ContactUsLinkComponent
RNA isolation, total RNA kit, E-Z 96®
RNA isolation, total RNA kit, E-Z 96®
Supplier:  OMEGA BIO-TEK
Certificates
RNA isolation, total RNA kit, E-Z 96®
Supplier:  OMEGA BIO-TEK

Specifications

  • Method / Format:
    Multiwell plate
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Sample size:
    5×106 cells or 10 mg tissue
  • Description:
    E-Z 96® Total RNA Kit I

Specifications

Related Information

Frequently Bought Together