Order Entry
Switzerland
ContactUsLinkComponent
Double spatula, micro
Double spatula, micro
Catalog # USBE3303
undefined
Double spatula, micro
Catalog # USBE3303

Specifications

  • Length:
    185 mm
  • Blade width:
    5 mm
  • Pk:
    1

Specifications

About this item

Stainless steel.

  • Rigid
  • One spatula end (width of blade 5 mm)
  • One spoon end (width of spoon 5 mm)