Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Spoons
Spoon/raspatory, Aesculap®
Spoon/raspatory, Aesculap®
Spoon/raspatory, Aesculap®
Catalog # AESCDO612R
Supplier:
Aesculap
Expand image
Spoon/raspatory, Aesculap®
Catalog # AESCDO612R
Supplier:
Aesculap
Specifications
Description:
Sharp spoon/raspatory, Partsch, No. 2, curved
Length:
150 mm
Pk:
1
Spoon size:
4,6 mm
Specifications
About this item
Product Details
Product Details