Anti-Bach1 Rabbit Polyclonal Antibody
SERTAHP1426
: Serotec
- Antibody type:Primary
- Antigen symbol:BACH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Chimpanzee,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:liquid
- Storage buffer:PBS
- Storage temperature:+4 °C, –20 °C if preferred
- Shipping temperature:4 °C
- Size:50 µg
- Pk:50 µG
Type: Primary
Antigen: Bach1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Chimpanzee, Mouse, Rat