Anti-CD62P (P-Selectin) Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: Psel.KO.2.7]
SERTMCA2419A488
: Serotec
- Antibody type:Primary
- Antigen name:P-Selectin
- Antigen symbol:CD62P
- Clonality:Monoclonal
- Clone:Psel.KO.2.7
- Conjugation:Alexa Fluor® 488
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Horse,Sheep,Goat,Human,Bovine,Rat,Mouse,Cat
- Cross adsorption:No
- Format:liquid
- Storage buffer:PBS
- Storage temperature:+4 °C, –20 °C if preferred
- Concentration:0,05 mg/ml
- Shipping temperature:4 °C
- Immunogen:P-selectin transfected 300.19 cells.
- Purification:Purified by affinity chromatography on Protein G from tissue culture supernatant
- Size:100 Tests / 1 ml
- Pk:100 Tests
Type: Primary
Antigen: CD62P (P-Selectin)
Clonality: Monoclonal
Clone: Psel.KO.2.7
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine, Cat, Goat, Horse, Mouse, Rat, Sheep