Anti-CD11B (Integrin Alpha M Chain, Mac-1) Mouse Monoclonal Antibody [clone: CA16.3E10]
Catalog # SERTMCA1777S
Supplier: Serotec
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Integrin Alpha M
- Antigen-Symbol:ITGAM
- Klonalität:Monoklonal
- Klon:CA16.3E10
- Konjugation:Unconjugated
- Durchflusszytometrie:Yes
- Wirt:Mouse
- ImmunoChemie:Yes
- Immunpräzipitation:Yes
- Isotyp:IgG1
- Reaktivität:Pig,Mustelid,Goat,Beluga,Bovine,Dog,Mink,Cat
- Kreuzadsorption:No
- Format:liquid
- Lagerungspuffer:PBS
- Lagertemperatur:+4 °C, –20 °C if preferred
- Transporttemperatur:4 °C
- Größe:2 ml
- VE:2 mL
Specifications
About this item
Type: Primary
Antigen: CD11B (Integrin Alpha M Chain, Mac-1)
Clonality: Monoclonal
Clone: CA16.3E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Beluga whale, Bovine, Cat, Goat, Mink, Mustelid, Pig