Anti-CD3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CA17.2A12]
SERTMCA1774F
: Serotec
- Antibody type:Primary
- Antigen symbol:CD3
- Clonality:Monoclonal
- Clone:CA17.2A12
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Dog
- Cross adsorption:No
- Format:liquid
- Storage buffer:PBS
- Storage temperature:+4 °C
- Concentration:0,1 mg/ml
- Shipping temperature:4 °C
- Purification:Purified by affinity chromatography on Protein G
- Size:100 μg
- Pk:0,1 mg
Do not freeze!
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: CA17.2A12
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog