Anti-Eosinophil Major Basic Protein Mouse Monoclonal Antibody [clone: BMK-13]
Catalog # SERTMCA5751
Supplier: Serotec
Specifications
- Antibody type:Primary
- Antigen name:Eosinophil Major Basic Protein
- Clonality:Monoclonal
- Clone:BMK-13
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Guinea pig,Rat
- Cross adsorption:No
- Format:liquid
- Storage buffer:PBS
- Storage temperature:+4 °C, –20 °C if preferred
- Concentration:0,1 mg/ml
- Shipping temperature:4 °C
- Size:100 μg
- Pk:1
Specifications
About this item
Type: Primary
Antigen: Eosinophil Major Basic Protein
Clonality: Monoclonal
Clone: BMK-13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Guinea Pig, Rat