ContactUsLinkComponent
Microplate sealing tape
Microplate sealing tape
Catalog # GENO786-422
Supplier:  G-Biosciences
undefined
Microplate sealing tape
Catalog # GENO786-422
Supplier:  G-Biosciences
Specifications
  • Description:
    Self-adhesive sealing tape, for 96- and 384-well plates, 100 sheets
  • Pk:
    1 KIT
Specifications
About this item
Acetate film with acrylic adhesive. This self-adhesive microplate sealing tape is suitable for sealing both 96-well and 384-well plates. The precut, pressure-senstive tape seals and reseals the surface of the plates during incubations to prevent evaporation and allow more vigorous mixing. The tape will reseal the plates 2 to 3 times.
  • Non sterile
  • Transparent and non pierceable
  • Minimise evaporation and condensation
  • For short and long term storage of plates
  • Temperature range: −16 to +38 °C