Anti-Alpha/ Beta-Synuclein Rabbit Polyclonal Antibody
Catalog # ABFRLF-PA0228
Supplier: AbFrontier
Specifications
- Antibody type:Primary
- Antigen name:Synuclein alpha/beta
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage temperature:Store for 1 year at –20 °C from date of shipment
- Concentration:1 mg/ml
- Immunogen:Recombinant human protein purified from E.coli (His-alpha-synuclein)
- Purification:Protein A purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Alpha/ Beta-Synuclein
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human