Order Entry
Switzerland
ContactUsLinkComponent
Racks for test tubes
Racks for test tubes
Catalog # HACH2587300
Supplier:  Hach
undefined
Racks for test tubes
Catalog # HACH2587300
Supplier:  Hach

Specifications

  • For tube Ø:
    10 - 13 mm
  • No. of Holes:
    90 (6×15)
  • Material:
    PP
  • W×D×H:
    248×105×70 mm
  • Pk:
    1

Specifications

About this item

PP, no-wire.

  • Numbered and lettered rows aid in identification of tubes
  • Collapsible for storage but can be assembled in seconds

These racks feature a locking mechanism that prevents 'floaters' in water baths and also speeds decanting by allowing the whole rack to be inverted. Manufactured from PP that is more dense than water assures that the rack does not float when immersed.