ContactUsLinkComponent
Petri dishes, disposable, square, Nunc™ Lab-Tek™
Petri dishes, disposable, square, Nunc™ Lab-Tek™
Catalog # NUNC4021
   Low Manufacturing Impact
undefined
Petri dishes, disposable, square, Nunc™ Lab-Tek™
Catalog # NUNC4021
   Low Manufacturing Impact
Specifications
  • Sterility:
    Sterile
  • Version:
    Square, 100×15 mm, vented, without stacking ring
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    500
Specifications
About this item
Square Petri dishes for culturing fungi, bacteria and other microorganisms.
Packaging: 20 plates per sealed plastic sleeve.