ContactUsLinkComponent
Molecular sieve 3A (0.3 nm, 3 Å), BAKER ANALYZED® 8-12 Mesh, activated, J.T.Baker®
Molecular sieve 3A (0.3 nm, 3 Å), BAKER ANALYZED® 8-12 Mesh, activated, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Molecular sieve 3A (0.3 nm, 3 Å), BAKER ANALYZED® 8-12 Mesh, activated, J.T.Baker®
Supplier:  Avantor
Certificates
Specification Test Results
  • For Laboratory,Research,or Manufacturing Use:
    = 5 %
  • Loss on Drying at 150°C:
    ≤ 5 %
  • Mesh - Thru U.S. No. 12 Sieve:
    = 20 %