Orders LinkContactUsLinkComponent
High leverage end cutter
High leverage end cutter
High leverage end cutter, chrome plated, handles insulated with multi-component grips
Catalog # 238-5330
Supplier:   KNIPEX WERKE
High leverage end cutter
Catalog # 238-5330
Supplier:  KNIPEX WERKE
High leverage end cutter
High leverage end cutter, chrome plated, handles insulated with multi-component grips
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Chrome vanadium heavy-duty steel, forged, oil-hardened. For heaviest duty. Chrome plated head. With two-coloured insulated multi-component grips.
  • High cutting performance with minimum effort
  • Suitable for soft, hard and piano wire
  • Precision cutting edges additionally induction-hardened
Length: 160 mm
Specifications
  • Description:
    High leverage end cutter, chrome plated, handles insulated with multi-component grips