Bottles, narrow neck, Square, PC, without screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PC, transparent, without screw cap.
- Easy-to-handle square design
- Tough, durable even at low temperatures
- With molded-in graduations and drip-proof neck
Ideal for storage of aqueous solutions and short-term storage of media.
These bottles are autoclavable.
Specifications
- Catalog No:
- NALG362015-0250
- NALG362015-0125
- 215-7000
- NALG362015-0500
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement