Safety wash bottles, wide neck, multilingual, Azlon®
Supplier: SciLabware
Certificates
About this item
LDPE, translucent, with PP spray cap.
- Indelibly printed in four languages (English, French, German and Spanish)
- Solvent labels including the new Globally Harmonised Symbols, NFPA diamond, risk phrases and safety information
- Closure feature precision jet tip aperture for fine control
Wide neck bottles fitted with colour coded one-piece PP closure that maintain their shape in use.
Certifications: Azlon® wash bottles meet the material requirements of the U.S. Food and Drug Administration as specified in 21 CFR 177.1520.
Frequently Bought Together