Bottles, narrow neck, round, HDPE, with thread
Nome del fornitore: EP SCIENTIFIC
Certificati
Informazioni su questo articolo
HDPE, translucent.
- Narrow neck for precision pouring
- Slim bottle design for easy grip and handling
- For general purpose sampling where cleaning and certification is not required or ready for own cleaning and certification procedures
Narrow neck round bottle are general purpose containers which are unprocessed and uncertified.
Ordering information: Bulk packed, caps must be ordered separately.
Specifiche
- Catalog No:
- EPSC013-1000BP
- EPSC013-0250BP
- EPSC013-0500BP
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Specifiche
Frequently Bought Together