Anti-TFPI Mouse Monoclonal Antibody [clone: 2E5]
Catalog # ABNOH00007035-M01A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Tissue Factor Pathway Inhibitor
- Antigen symbol:TFPI
- Clonality:Monoclonal
- Clone:2E5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Form:Ascites fluid
- Gene ID:7035
- Antigen synonyms:TFI|EPI|lipoprotein-associated coagulation inhibitor|LACI|tissue factor pathway inhibitor|anti-convertin|TFPI1|extrinsic pathway inhibitor
- Amino acid number:1 to 251
- Sequence:MIYTMKKVHALWASVCLLLNLAPAPLNADSEEDEEHTIITDTELPPLKLMHSFCAFKADDGPCKAIMKRFFFNIFTRQCEEFIYGGCEGNQNRFESLEECKKMCTRDNANRIIKTTLQQEKPDFCFLEEDPGICRGYITRYFYNNQTKQCERFKYGGCLGNMNNFETLEECKNICEDGPNGFQVDNYGTQLNAVNNSLTPQSTKVPSLFVTKEGTNDGWKNAAHIYQVFLNAFCIHASMFFLGLDSISCLC
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:TFPI (AAH15514, 1 a.a. ~ 251 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:200 μl
- Pk:200 µl
Specifications
About this item
Mouse monoclonal antibody raised against a full-length recombinant TFPI.
Type: Primary
Antigen: TFPI
Clonality: Monoclonal
Clone: 2E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human