Anti-TFPI Mouse Monoclonal Antibody [clone: AHTFPI-5138]
Catalog # ABNOMAB0281
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Tissue Factor Pathway Inhibitor
- Antigen symbol:TFPI
- Clonality:Monoclonal
- Clone:AHTFPI-5138
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:7035
- Antigen synonyms:TFI|EPI|lipoprotein-associated coagulation inhibitor|LACI|tissue factor pathway inhibitor|anti-convertin|TFPI1|extrinsic pathway inhibitor
- Storage buffer:PBS (50% glycerol)
- Sequence:Asp-Ser-Glu-Glu-Asp-Glu-Glu-His-Thr-Ile-Ile-Thr-Asp-Thr-Glu-Cys
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 16 amino acids at N-terminus of human TFPI.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against synthetic peptide of TFPI.
Recommended Dilutions: Western Blot: 5 ?g/ml
Type: Primary
Antigen: TFPI
Clonality: Monoclonal
Clone: AHTFPI-5138
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human