Anti-AMPD1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB22103
Anbieter: Abnova
Technische Daten
- Antibody type:Primary
- Antigen name:AMP deaminase 1
- Antigen symbol:AMPD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:270
- Antigen synonyms:Ampd1|Myoadenylate deaminase|Ampd-1|AMP deaminase isoform M|AI553520
- Storage buffer:PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
- Sequence:AEEKQIDDAMRNFAEKVFASEVKDEGGRQEISPFDVDEICPISHHEMQAHIFHLETLSTSTEARRKKRFQGRKTVNLSIPL
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human AMPD1.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Technische Daten
Über diesen Artikel
Rabbit polyclonal antibody raised against recombinant AMPD1.
Recommended Dilutions: Immunohistochemistry: 1:20-1:50
Type: Primary
Antigen: AMPD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human