Anti-MTOR Rabbit Polyclonal Antibody
Katalognr. ABNOPAB10335
Anbieter: Abnova
Technische Daten
- Antibody type:Primary
- Antigen name:Mechanistic Target of Rapamycin
- Antigen symbol:MTOR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:2475
- Antigen synonyms:FRAP1|mTOR|m-TOR|RAPT1|RAFT1|FKBP-Rapamycin Associated Protein|Mechanistic Target Of Rapamycin|EC 2.7.11.1|FRAP|FRAP2|Rapamycin Target Protein|FK506 binding protein 12-rapamycin associated protein|Mammalian Target Of Rapamycin|Serine/Threonine-Protein Kinase MTOR|MTOR
- Modification:phospho-Ser2448
- Storage buffer:20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding S2448 of human MTOR.
- Size:100 μg
- Pk:100 µg
Technische Daten
Über diesen Artikel
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MTOR.
Recommended Dilutions: ELISA: 1:15000-1:65000; Western Blot: 1:500-1:2000; Immunohistochemistry: 5.0 ?g/ml
Type: Primary
Antigen: MTOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human