Anti-MAT2A Mouse Monoclonal Antibody [clone: AT3A2]
Catalog # ABNOMAB3213
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Methionine Adenosyltransferase 2 Alpha
- Antigen symbol:MAT2A
- Clonality:Monoclonal
- Clone:AT3A2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:4144
- Antigen synonyms:SAMS2|MATA2|MATII
- Storage buffer:PBS, pH 7,4 (0,1% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human MAT2A.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant MAT2A.
Type: Primary
Antigen: MAT2A
Clonality: Monoclonal
Clone: AT3A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human