Anti-MAT2A Rabbit Polyclonal Antibody
Catalog # ABNOPAB12388
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Methionine Adenosyltransferase 2 Alpha
- Antigen symbol:MAT2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Primates,Pig,Chicken,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:4144
- Antigen synonyms:SAMS2|MATA2|MATII
- Amino acid number:1 to 100
- Storage buffer:PBS (30% glycerol, 0,1% sodium azide)
- Storage temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 1-100 of human MAT2A.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of MAT2A.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: MAT2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Human, Mouse, Pig, Primates, Rat