Anti-MOMP Mouse Monoclonal Antibody [clone: CL13-227.1.2]
Catalog # ABNOMAB6161
Anbieter: Abnova
Technische Daten
- Antibody type:Primary
- Antigen name:Major Outer Membrane Protein
- Antigen symbol:MOMP
- Clonality:Monoclonal
- Clone:CL13-227.1.2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:C. trachomatis
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,4
- Storage temperature:Store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Chlamydia trachomatis elementary bodies.
- Pk:100 µG
Technische Daten
Über diesen Artikel
Mouse monoclonal antibody raised against Chlamydia trachomatis MOMP.
Type: Primary
Antigen: MOMP
Clonality: Monoclonal
Clone: CL13-227.1.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: C. trachomatis