Anti-CLIC4 Goat Polyclonal Antibody
Catalog # ABNOPAB7536
Anbieter: Abnova
Technische Daten
- Antibody type:Primary
- Antigen name:Chloride Intracellular Channel 4
- Antigen symbol:CLIC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:25932
- Antigen synonyms:H1|huH1|p64H1|CLIC4L|MTCLIC
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:NGLKEEDKEPLIE-C
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human CLIC4.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Technische Daten
Über diesen Artikel
Goat polyclonal antibody raised against synthetic peptide of CLIC4.
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.1-0.3 ?g/ml; Immunofluorescence: 5 ?g/ml
Type: Primary
Antigen: CLIC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse