Order Entry
Switzerland
ContactUsLinkComponent
Mouse IgG2b Isotype Control [clone: MPC-11] (redFluor® 710)
Mouse IgG2b Isotype Control [clone: MPC-11] (redFluor® 710)
Catalog # 80-4732-U100
Supplier:  Tonbo Biosciences
Mouse IgG2b Isotype Control [clone: MPC-11] (redFluor® 710)
Catalog # 80-4732-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Control
  • Clone:
    MPC-11
  • Conjugation:
    redFluor® 710
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Neutralisation:
    Yes
  • Storage temperature:
    Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.

Type: Control
Antigen: MPC-11
Clonality:
Clone: MPC-11
Conjugation: redFluor™ 710
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: N/A