ContactUsLinkComponent
Cryo storage box Cryo-Safe™ Cold Box, SP Bel-Art
Cryo storage box Cryo-Safe™ Cold Box, SP Bel-Art
Catalog # 479-1150
undefined
Cryo storage box Cryo-Safe™ Cold Box, SP Bel-Art
Catalog # 479-1150
Specifications
  • For:
    12 microcentrifuge tubes 1,5 ml
  • Dimensions:
    117×117×71 mm
  • Pk:
    1
Specifications
About this item
Polystyrene box with polystyrene cover.
  • Practical cold storage of perishable samples
  • Capacity: twelve 1,5 ml microcentrifuge tubes (tubes not included)
  • Holds −15 ºC temperature for 1,5 hours, and maintains 0 ºC for 4 hours when removed from a freezer
The Cryo-Safe™ Cold Box safeguards enzymes and thermolabile substances from thermal degradation. This compact unit is made up of a molded polystyrene box with cover. The interior of the unit is filled with a cryogenic compound and then permanently sealed, to ensure the low temperatures needed to safeguard materials in the cold box.