These syringe filters are ideal for filtration, clarification or particle removal from buffers, reagents, DMSO, HPLC/GC solvents.
- Sterile filters are certified sterile, non-cytotoxic and non-pyrogenic
- Accommodate sample volumes from 10 to 100 ml
SFCA membrane; PP housing: Use for immunological samples and biological fluids. Low protein binding, lower extractables than standard CA.
CA membrane; PP housing: Ideal for buffers, aqueous reagents and solutions, including EIA and ELISA samples. Some protein binding and extractables. For sterile filtration and clarification of buffers, aqueous solutions and reagents.
Nylon membrane; PP housing: Good for cryoprotectants such as DMSO and filtration of aqueous biological solutions, alcoholic solutions and non-aqueous HPLC/GC solvents. Naturally hydrophilic, low extractables, some protein binding.
PTFE membrane; PP housing: Naturally hydrophobic for sterile venting applications and filtration of samples in HPLC/GC solvents. Ideal for sterile venting of air or other gases during filtration.
Glass fibre membrane; PP housing: These syringe filters are designed for filtering high particulate or viscous suspensions or solutions. Disposable pre-filter. Prevents clogging of final filter. Increases sample recovery.
- Catalog No:
- 514-0455
- 513-1712
- 516-0932
- 516-0933
- 513-1762
- 516-0931
- 513-1761
- 516-0949
- 516-0947
- 516-0948
- 516-0942
- 516-0950
- 516-0940
- 516-0941
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- IRECS
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement