Anti-ITGA2 Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: ]
Type: Primary
Antigen: ITGA2
Clonality: Monoclonal
Clone:
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Mouse
- Catalog No:
- BLEG108921
- BLEG108922
- Antigen symbol:
- ITGA2
- ITGA2
- Antigen name:
- Integrin Alpha 2
- Integrin Alpha 2
- Antigen synonyms:
- BR|GPIa|VLAA2|DX5 antigen|pan-NK cells|CD49B|HPA-5|VLA-2|ITGA2
- BR|GPIa|VLAA2|DX5 antigen|pan-NK cells|CD49B|HPA-5|VLA-2|ITGA2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)-Cy7®
- PE (Phycoerythrin)-Cy7®
- Reactivity:
- Baboon, Capuchin monkey, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Squirrel monkey
- Baboon, Capuchin monkey, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Squirrel monkey
- Host:
- Rat
- Rat
- Isotype:
- IgM
- IgM
- Flow cytometry:
- Yes
- Yes
Frequently Bought Together