Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD69 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ]
Anti-CD69 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ]
Supplier:  Biolegend
Certificates
Anti-CD69 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • BLEG310910
  • BLEG310909
  • Antigen symbol:
  • CD69
  • CD69
  • Antigen name:
  • CD69 molecule
  • CD69 molecule
  • Antigen synonyms:
  • EA1|MLR-3|AIM|GP32/28|CLEC2C|BL-AC/P26
  • EA1|MLR-3|AIM|GP32/28|CLEC2C|BL-AC/P26
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Reactivity:
  • Marmoset, African green monkey, Chimpanzee, Human, Squirrel monkey
  • Marmoset, African green monkey, Chimpanzee, Human, Squirrel monkey
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes

Specifications

Frequently Bought Together