Anti-TBX21 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 4B10]
Supplier: Biolegend
Certificates
About this item
Type: Primary
Antigen: TBX21
Clonality: Monoclonal
Clone:
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Specifications
- Catalog No:
- BLEG644815
- BLEG644816
- Antigen symbol:
- TBX21
- TBX21
- Antigen name:
- T-box 21
- T-box 21
- Antigen synonyms:
- T-box transcription factor TBX21|TBX21 |T-cell-specific T-box transcription factor T-bet|T-PET|T-box protein 21|T-bet|TBLYM|TBET
- T-box transcription factor TBX21|TBX21 |T-cell-specific T-box transcription factor T-bet|T-PET|T-box protein 21|T-bet|TBLYM|TBET
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Brilliant Violet® 421
- Brilliant Violet® 421
- Clone:
- 4B10
- Reactivity:
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 30009
- Isotype:
- IgG1
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421® under optimal conditions. The solution is free of unconjugated Brilliant Violet 421® and unconjugated antibody.
- Storage temperature:
- The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:
- Ambient RT
Specifications
Frequently Bought Together