Anti-IL17F Goat Polyclonal Antibody (Alexa Fluor® 488) [clone: ]
Nome del fornitore: Biolegend
Certificati
Informazioni su questo articolo
Type: Primary
Antigen: IL17F
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Specifiche
- Catalog No:
- BLEG516603
- BLEG516604
- Antigen symbol:
- IL17F
- IL17F
- Antigen name:
- Interleukin F
- Interleukin F
- Antigen synonyms:
- cytokine ML-1|mutant IL-17F|IL-17F|CANDF6|interleukin-17F|ML-1|interleukin-24|IL-24|ML1
- cytokine ML-1|mutant IL-17F|IL-17F|CANDF6|interleukin-17F|ML-1|interleukin-24|IL-24|ML1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Alexa Fluor® 488
- Alexa Fluor® 488
- Reactivity:
- Mouse
- Mouse
- Host:
- Goat
- Goat
- Flow cytometry:
- Yes
- Yes
Specifiche
Frequently Bought Together