Anti-ITGAX Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ]
Supplier: Biolegend
Certificates
About this item
Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- BLEG337207
- BLEG337208
- Antigen symbol:
- ITGAX
- ITGAX
- Antigen name:
- Integrin Subunit Alpha X
- Integrin Subunit Alpha X
- Antigen synonyms:
- CD11C|SLEB6
- CD11C|SLEB6
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Reactivity:
- Mouse
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
Specifications
Frequently Bought Together