Anti-PTPRC Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: A20]
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: A20
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse
- Catalog No:
- BLEG110732
- BLEG110731
- Antigen symbol:
- PTPRC
- CD45.1
- Antigen name:
- Protein tyrosine phosphatase receptor type C
- CD45.1 molecule
- Antigen synonyms:
- CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Brilliant Violet® 421
- Brilliant Violet® 421
- Clone:
- A20
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 19264
- Isotype:
- IgG2a kappa
- IgG2a
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Immunogen:
- SJL mouse thymocytes and splenocytes
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421® under optimal conditions. The solution is free of unconjugated Brilliant Violet 421® and unconjugated antibody.
- Concentration:
- 0,2 mg/ml
- Storage temperature:
- The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:
- Ambient RT
Frequently Bought Together