Anti-CB1 Rabbit Polyclonal Antibody
Catalog # CAYM10006590-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:CB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Amino acid number:461 to 472
- Sequence:MSVSTDTSAEAL
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human CB1 receptor amino acids 461-472
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: CB1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat