Anti-SCN10A Mouse Monoclonal Antibody (Unconjugated) [clone: S134-12]
Katalognr. ROCK200-301-G22
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:sodium channel, voltage-gated, type X, alpha subunit
- Antigen symbol:SCN10A
- Clonality:Monoclonal
- Clone:S134-12
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:29571
- Antigen synonyms:Nav1.8|Na(V)1.8|PN3
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Nav1.8 Antibody was produced in mice by repeated immunizations raised against a fusion Protein corresponding near the cytoplamsic C-terminus region of rat Nav1.8.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-NAV1.8 Antibody is suitable for use in WB, IHC and IF microscopy. Expect a band approximately ~220kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: Immunofluorescence: 1.0-10µg/ml; Western Blot: 1-10µg/ml; Immunochemistry: 0.1-1.0µg/ml
Type: Primary
Antigen: SCN10A
Clonality: Monoclonal
Clone: S134-12
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat