Anti-Protein Gene Product 9.5 Mouse Monoclonal Antibody [clone: 13C4]
Supplier: US Biological
Certificates
About this item
Recommended dilution: Immunofluorescence: 1-2?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: Protein Gene Product 9.5
Clonality: Monoclonal
Clone: 13C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Bovine, Dog, Guinea pig, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish,
Specifications
- Catalog No:
- USBI138590
- USBI138590-100
- Antigen name:
- Protein Gene Product 9.5
- Protein Gene Product 9.5
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 13C4
- 13C4
- Reactivity:
- Zebrafish, Pig, Rabbit, Sheep, Bovine, Guinea pig, Dog, Rat, Mouse
- Zebrafish, Pig, Rabbit, Sheep, Bovine, Guinea pig, Dog, Rat, Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- PGP9.5 protein from human brain
- PGP9.5 protein from human brain
- Form:
- liquid
- liquid
- Purification:
- purified by Protein G chromatography
- purified by Protein G chromatography
- Concentration:
- ~0,2 mg/ml
- ~0,2 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, 0,05% BSA, 0,05% sodium azide.
- PBS, 0,05% BSA, 0,05% sodium azide.
- Storage temperature:
- –20 °C
- –20 °C
Specifications
Frequently Bought Together