Anti-Protein Gene Product 9.5 Mouse Monoclonal Antibody [clone: 5G201]
Catalog # USBIP9102-70D-250
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Protein Gene Product 9.5
- Clonality:Monoclonal
- Clone:5G201
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Epitope:full-length
- Cross adsorption:No
- Format:Cell culture supernatant
- Form:liquid
- Storage temperature:–20 °C
- Immunogen:Recombinant protein corresponding to full length human protein gene product 9.5 molecule.
- Size:250 μl
- Pk:250 µl
Specifications
About this item
Type: Primary
Antigen: Protein Gene Product 9.5
Clonality: Monoclonal
Clone: 5G201
Conjugation: unconjugated
Epitope: full-length
Host: Mouse
Isotype: IgG2b
Reactivity: Human,