Anti-Calcitonin Gene Related Peptide Mouse Monoclonal Antibody [clone: 2Q2268]
Catalog # USBIC0115-01C-100
Anbieter: US Biological
Technische Daten
- Antibody type:Primary
- Antigen name:Calcitonin Gene Related Peptide
- Antigen symbol:CGRP
- Clonality:Monoclonal
- Clone:2Q2268
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Guinea pig
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, 1% BSA and 0,1% sodium azide.
- Storage temperature:–20 °C
- Concentration:~0,1 mg/ml
- Immunogen:A terminal fragment of CGRP-coupled to BSA with glutaraldehyde
- Purification:purified by Protein G chromatography
- Size:100 μg
- Pk:100 µG
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: Calcitonin Gene Related Peptide
Clonality: Monoclonal
Clone: 2Q2268
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Guinea pig, Human,