Anti-Thyroid Hormone Receptor Beta Mouse Monoclonal Antibody [clone: SPM222]
GNTXGTX17898
: Genetex
- Antibody type:Primary
- Antigen name:Thyroid Hormone Receptor Beta
- Clonality:Monoclonal
- Clone:SPM222
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Dog
- Amino acid number:31 to 50
- Storage temperature:+4 °C
- Immunogen:Synthetic peptide: GMSEACLHRKSHSERRSTLK, corresponding to N terminal amino acids 31-50 of Human Thyroid Hormone Receptor beta.
- Purification:purified by Protein G chromatography
- Pk:250 µl
Type: Primary
Antigen: Thyroid Hormone Receptor Beta
Clonality: Monoclonal
Clone: [SPM222]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog