Anti-TDT Mouse Monoclonal Antibody [clone: HT-1, HT-4, HT-8, HT-9 (FITC)]
GNTXGTX43336
: Genetex
- Antibody type:Primary
- Antigen symbol:TDT
- Clonality:Monoclonal
- Clone:HT-1, HT-4, HT-8, HT-9 (FITC)
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Reactivity:Human
- Storage temperature:–20 °C
- Size:25 µg
- Pk:25 µG
Type: Primary
Antigen: TDT
Clonality: Monoclonal
Clone: [HT-1, HT-4, HT-8, HT-9] (FITC)
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1/IgG2a
Reactivity: Human