Anti-S100 Mouse Monoclonal Antibody [clone: 3B10]
GNTXGTX28334
: Genetex
- Antibody type:Primary
- Antigen symbol:S100
- Clonality:Monoclonal
- Clone:3B10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Storage temperature:+4 °C
- Immunogen:Full length protein (Human)
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: S100
Clonality: Monoclonal
Clone: [3B10]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human