Anti-IL1 beta Mouse Monoclonal Antibody [clone: 5C10]
Catalog # GNTXGTX28321
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 1 beta
- Antigen symbol:IL1B
- Clonality:Monoclonal
- Clone:5C10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgM
- Reactivity:Human
- Storage temperature:+4 °C
- Immunogen:Recombinant beta human Interleukin-1
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: IL1 beta
Clonality: Monoclonal
Clone: [5C10]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human