Order Entry
Switzerland
ContactUsLinkComponent
Anti-HLA CLASS I Mouse Monoclonal Antibody [clone: MEM-147]
Anti-HLA CLASS I Mouse Monoclonal Antibody [clone: MEM-147]
Catalog # GNTXGTX29090
Supplier:  Genetex
Anti-HLA CLASS I Mouse Monoclonal Antibody [clone: MEM-147]
Catalog # GNTXGTX29090
Supplier:  Genetex

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HLA Class 1
  • Clonality:
    Monoclonal
  • Clone:
    MEM-147
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    –20 °C
  • Immunogen:
    PHA-activated peripheral blood lymphocytes
  • Purification:
    95% by SDS-PAGE
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: HLA CLASS I
Clonality: Monoclonal
Clone: [MEM-147]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human