Anti-E2F1 Mouse Monoclonal Antibody [clone: KH20]
Catalog # GNTXGTX23221
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen symbol:E2F1
- Clonality:Monoclonal
- Clone:KH20
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Storage temperature:+4 °C
- Immunogen:Recombinant full length protein (Human).
- Purification:purified by Protein A chromatography
- Pk:500 µl
Specifications
About this item
Type: Primary
Antigen: E2F1
Clonality: Monoclonal
Clone: [KH20]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human