Anti-RNP M3-M4 Mouse Monoclonal Antibody [clone: 2A6]
Catalog # GNTXGTX30273
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen symbol:RNP M3-M4
- Clonality:Monoclonal
- Clone:2A6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Format:Ascites fluid
- Storage temperature:–70 °C
- Immunogen:M19 fusion protein containing full-length human protein.
- Purification:Ascitic fluid
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: RNP M3-M4
Clonality: Monoclonal
Clone: [2A6]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human