Anti-CD14 Mouse Monoclonal Antibody [clone: MEM-18]
Catalog # GNTXGTX28103
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen symbol:CD14
- Clonality:Monoclonal
- Clone:MEM-18
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Storage temperature:–20 °C
- Immunogen:A crude mixture of human urinary proteins precipitated by ammonium sulphate from the urine of a patient suffering from proteinuria.
- Purification:95% by SDS-PAGE
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: [MEM-18]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human